Learn More
Purell™ Professional Advanced Hand Sanitizer Fragrance Free Gel
PURELL™ Professional Advanced Hand Sanitizer Fragrance Free Gel for PURELL™ Hand Sanitizer Dispensers.
Manufacturer:  Gojo Industries IncSupplier Diversity Partner 646002
 View more versions of this product
Catalog No. 19-043-802
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Green certified
  • Fragrance-free gel hand sanitizer for professional environments
  • Kills more than 99.99% of most common germs that may cause illness
  • Outperforms other hand sanitizers ounce for ounce
  • Made with naturally renewable ethanol
  • Clinically proven formulation to maintain skin health
  • Dye and fragrance free
  • Supports LEED certification criteria
  • AT-A-GLANCE™ refills - Monitor product level with just one look
  • SANITARY SEALED™ PET refill is easily recycled
Specifications
Specifications
Safety
2
Hand Sanitizer
PURELL™ ES6 Touch-Free Hand Sanitizer Dispensers
40.577 fl oz
Clear
Fragrance Free
1200 ml