Learn More
PDI™ Povidone-Iodine Prep Pad
Provides proven antisepsis
Supplier:  PDI™ C12400
Catalog No. 06-669-98
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
  • Non-irritating and virtually non-stinging for better patient comfort
  • Perfect for venipuncture, I.V. starts, kidney dialysis, pre-op prep, and other minor invasive procedures
  • Saturated with a 10% USP povidone-iodine prep solution provide proven antisepsis
Order Info
Pad size: 1.75 x 3.5 in. (4.4 x 8.8cm). 100 individually wrapped pads per box, 10 boxes per case.
Specifications
Specifications
PVP Iodine Prep Pad
For venipuncture, I.V. starts, kidney dialysis, pre-op prep, and other minor invasive procedures, provides longer germicidal activity than ordinary Iodine solution
Recommendations
Recommendations