Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEDTOX™ PROFILE-II ER™ Cassette Drug Testing Device for AMP, BAR, BZO, COC, MAMP (MDMA), MTD, OPI, OXY, PCP, PPX, TCA, TH
Click to view available options
Quantity:
25 Pack
Description
Includes
Pipette
- Visual read, single-assay devices deliver accurate screening results in minutes
- 510(k)-cleared
- Made in the USA
- Quality management system conforms with ISO 13485:2003
- Accurate screening results in minutes
- On-board adulteration strip available on some products
- Tests for up to 12 drugs of abuse in a single device
- Technical support services available for all MEDTOX Diagnostics, Inc. products
Amphetamine (AMP): 1000ng/mL
Barbiturates (BAR): 200ng/mL
Benzodiazepine(BZO): 300ng/mL
Cocaine (COC): 300ng/mL
3,4-Methylenedioxy-N-Methylamphetamine (MDMA) 1500ng/mL
Methamphetamine (MAMP): 1000ng/mL
Methadone (MTD): 300ng/mL
Opiates (OPI3): 300ng/mL
Oxycodone (OXY): 100ng/mL
Phencyclidine (PCP): 25ng/mL
Tricyclic Antidepressant (TCA): 300ng/mL
Propoxyphene (PPX): 300ng/mL
Cannabinoids (THC): 50ng/mL
Specifications
Specifications
Sample Type | Urine |
For Use With (Application) | Twelve panel drugs of abuse test for AMP/BAR/BZO/COC/MAMP/MTD/OPI/OXY/PCP/PPX/TCA/THC - specimen type is urine |
Quantity | 25 Pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction