Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEDTOX™ PROFILE-II ER™ Cassette Drug Testing Device for AMP, BAR, BZO, COC, MAMP (MDMA), MTD, OPI3, PPX, TCA, THC
Conduct on-site screening for up to 12 drugs of abuse including Cannabinoids, Cocaine, Oxycodone, Methamphetamine and Opiates. MEDTOX™ Diagnostics PROFILE-II ER™ Cassette Drug Testing Devices are easy to use and cost-effective.
Supplier: MEDTOX™ 601636
Includes: Pipette
Description
- Visual read, single-assay devices deliver accurate screening results in minutes
- 510(k)-cleared
- Made in the USA
- Quality management system conforms with ISO 13485:2003
- Accurate screening results in minutes
- On-board adulteration strip available on some products
- Tests for up to 12 drugs of abuse in a single device
- Technical support services available for all MEDTOX Diagnostics, Inc. products
Amphetamine (AMP): 1000ng/mL
Barbiturates (BAR): 200ng/mL
Benzodiazepine(BZO): 300ng/mL
Cocaine (COC): 300ng/mL
3,4-Methylenedioxy-N-Methylamphetamine (MDMA) 1500ng/mL
Methamphetamine (MAMP): 1000ng/mL
Methadone (MTD): 300ng/mL
Opiates (OPI3): 300ng/mL
Oxycodone (OXY): 100ng/mL
Phencyclidine (PCP): 25ng/mL
Tricyclic Antidepressant (TCA): 300ng/mL
Propoxyphene (PPX): 300ng/mL
Cannabinoids (THC): 50ng/mL
Specifications
Urine | |
Ten panel drugs of abuse test for AMP/BAR/BZO/COC/MAMP/MTD/OPI3/PPX/TCA/THC - specimen type is urine | |
25 Pack |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction