Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Profilin 1 Rabbit anti-Human, Mouse, Rat, Bovine, DyLight™ 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP119344UV

Catalog No. NBP119344UV


Add to cart

Description

Description

Profilin 1 Polyclonal antibody specifically detects Profilin 1 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Profilin 1
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
profilin 1, Profilin I, profilin-1
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Bovine, Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
PFN1
Synthetic peptide made to a C-terminal portion of human Profilin 1 (within residues 100-140). [Swiss-ProtNo. P07737]
Immunogen affinity purified
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors
Primary
5216.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit