Learn More
Profilin 1 Mouse anti-Canine, Human, Non-human primate, Rat, Clone: OTI2D2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525120
Catalog No. PIMA525120
Add to cart
Description
Description
Profilin is an ubiquitous small (12-15 kDa) phosphoinositides and poly-L-proline binding protein that plays a role both in signal transduction pathways and actin filament dynamics. There are two mammalian profilins with similar biochemical properties but different expression patterns. Profilin I appears to be highly expressed in most tissues except for skeletal muscle, whereas profilin II is predomitly expressed in brain and at lower levels also in skeletal muscle, uterus and kidney. Profilin is a mainly cytosolic protein with higher concentrations in dynamic membrane areas like the leading edge and ruffling membranes.
Specifications
Specifications
Profilin 1
Monoclonal
0.58 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P62963, P07737
Epididymis tissue protein Li 184a, PFN1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5216, 607397, 64303, 710753
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2D2
Unconjugated
PFN1
Liquid
Pfn1
Full length human recombit protein of PFN1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate, Rat