Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ ProFlex™ 760 Impact-Reducing Utility Work Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- TPR deflects knuckle busting impacts and scrapes
- Tena-Grip™ reinforced palm, fingertips, and thumb saddle
- Technical hex-stretch knit construction
- Molded hook and loop closure
- Gloves feature ID space to write user's names
Specifications
Color | Black |
Hand | Specific |
Material | Thermoplastic Rubber |
Product Type | Work Gloves |
Size | 2X-Large |
For Use With (Application) | Heavy Duty Work, Construction, Mining, Oil and Gas, Mechanics, and Tool/Machine Operation |
Glove Cuff Style | Open |
Series | ProFlex™ 760 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction