Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 760 Impact-Reducing Utility Work Gloves

Thermoplastic rubber guards knuckles from impacts and scrapes, while padded palms are reinforced with Tena-Grip™ for additional protection.

Specifications

Color Black
For Use With (Application) Heavy Duty Work, Construction, Mining, Oil and Gas, Mechanics, and Tool/Machine Operation
Glove Cuff Style Open
Material Thermoplastic Rubber (TPR)
Product Type Work Gloves
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-820-187
View Documents
Ergodyne™
17662
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-820-188
View Documents
Ergodyne™
17663
Medium This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-820-189
View Documents
Ergodyne™
17664
Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-820-191
View Documents
Ergodyne™
17665
X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-820-192
View Documents
Ergodyne™
17666
2X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • TPR deflects knuckle busting impacts and scrapes
  • Tena-Grip™ reinforced palm, fingertips, and thumb saddle
  • Technical hex-stretch knit construction
  • Molded hook and loop closure
  • Gloves feature ID space to write user's names
Specifications

Specifications

Black
Open
Work Gloves
Heavy Duty Work, Construction, Mining, Oil and Gas, Mechanics, and Tool/Machine Operation
Thermoplastic Rubber (TPR)
ProFlex™ 760
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit