Learn More
Drg International IncSupplier Diversity Partner Progesterone Clia DFS Item
Manufacturer:  Drg International IncSupplier Diversity Partner CLA4663
A chemiluminescence immunoassay for the quantitative determination of progesterone in serum and plasma. The DRG Progesterone CLIA Kit is a chemiluminescence immunoassay (CLIA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal antibody directed towards an antigenic site on the Progesterone molecule. Endogenous Progesterone of a sample competes with a Progesterone horseradish peroxidase conjugate for binding to the coated antibody.
Catalog No. 50-194-7528