Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Progesterone Antibody (6-5E-10B) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Progesterone |
---|---|
Clone | 6-5E-10B |
Concentration | 1 mg/mL |
Dilution | Radioimmunoassay |
Applications | Radioimmune Assays (RIA) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25310301
|
Novus Biologicals
NBP2531030.1MG |
0.1 mg |
Each for $627.50
|
N/A | |||||
NB25310302
|
Novus Biologicals
NBP2531030.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Progesterone Monoclonal specifically detects Progesterone in All Species samples. It is validated for ELISA, Immunohistochemistry, Radioimmunoassay, N/A.Specifications
Progesterone | |
1 mg/mL | |
Radioimmune Assays (RIA) | |
Unconjugated | |
Mouse | |
All species | |
Pregn 4 ene 3 20 dione | |
IgG1 κ | |
Protein A or G purified | |
This MAb is specific for progesterone. It exhibits minimal cross reactivity with related compounds in ELISA. It reacts with Progesterone-11a-HMS-BSA: 100%; 5-beta-Pregnane-3,20-dione: 48%; 5-alpha-Pregnane-3,20-dione: 26.4%; 17-alpha-Hydroxyprogesterone: 2.5% and 20-alpha-Hydroxyprogesterone: 0.04%. Progesterone is a steroid hormone synthesized from the cholesterol derivative, pregnenolone, in the cortex of the adrenal gland. Progesterone is secreted by the corpus luteum and acts to prepare the endometrium for the implantation of a fertilized egg. During pregnancy, it is secreted by the placenta to prevent spontaneous abortion and to stimulate the development of mammary tissue to produce milk. Thus, progesterone plays a central role in the reproductive events associated with the establishment and maintenance of pregnancy. Luteinized theca cells of normal ovary secrete progesterone. The determination of progesterone concentrations in the body fluids is of great value for endocrinologica |
6-5E-10B | |
Radioimmunoassay | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS with No Preservative | |
Progesterone- 11a-hemisuccinate conjugated to bovine serum albumin (Pr11a-HMS-BSA) | |
Primary | |
Store at 4C. | |
314 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title