Learn More
Progesterone R/NR3C3 Mouse anti-Human, Alexa Fluor™ 532, Clone: PR501, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233369AF532
Catalog No. NP233369A32
Add to cart
Description
Description
Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Progesterone R/NR3C3
PR501
Western Blot
NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
This MAb is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81kDa) and PR-B (116kDa). Expression of PgR has been suggested to reflect a intact estrogen regulatory machinery and therefore, predict better clinical response to endocrine therapy than ER alone.
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
PGR
A progesterone receptor recombinant protein was used as the immunogen for the antibody. The specific epitope has not been mapped.
0.1 mL
Primary
5241
Human