Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Progesterone Receptor Ab-14, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-14, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS1882S1

 View more versions of this product

Catalog No. MS1882S1


Add to Cart

Description

Description

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: PgR636

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal (shared by A- and B-form)

Immunogen: Formalin fixed recombinant full-length A-form of human progesterone receptor

Molecular Weight: 116kDa (triplet, B-form) and 81kDa (singlet, A-form)

Positive Control: Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Progesterone Receptor Ab-14
Monoclonal
Unconjugated
Formalin fixed recombinant full length A-form of human progesterone receptor
500 μL
Cancer and Tumor Biology
Human
IgG2a κ
Immunohistochemistry (Paraffin)
PgR636
Mouse
Breast CA
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit