Learn More
Progesterone Receptor Ab-6 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-6, Mouse Monoclonal Antibody.
Supplier: Epredia MS196P1ABX
Description
Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.Host Species: Mouse
Clone: hPRa 6
Isotype: IgG2b
Species Reactivity: Human, Cow, Rabbit, and Chicken. Does not react with monkey. Others not known.
Epitope: Not determined
Immunogen: Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Molecular Weight: 116kDa (triplet, B-form)
Positive Control: T47D cells
Cellular Localization: Nuclear Ab-6 preferentially reacts with B-form of progesterone receptor.
Recommended for:
- Western Blotting (Not optimum)
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Sedimentation in Sucrose Gradient
Note:
USA: RUO; Int'l: RUO
Specifications
Progesterone Receptor Ab-6 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Cancer and Tumor Biology | |
Bovine, Chicken, Human, Rabbit | |
IgG2b |
Immunofluorescence, Immunoprecipitation | |
hPRa 6 | |
Unconjugated | |
Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.