Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Progesterone Receptor Ab-8, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-8, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS298R7A

 View more versions of this product

Catalog No. MS298R7


Add to cart

Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Progesterone Receptor Ab-8, Mouse Monoclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 2 + hPRa 3

Isotype: IgG1/κ + IgG1/κ

Species Reactivity: Human, Horse, Sheep and Pig. Others not known.

Epitope: N-terminal half of human PgR

Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (B-form) and 81kDa (A-form)

Positive Control: T47D or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Sedimentation in Sucrose Gradient
Specifications

Specifications

Progesterone Receptor Ab-8
Monoclonal
Unconjugated
Prediluted
PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
7mL
Cancer and Tumor Biology
Primary
Equine, Human, Porcine, Sheep
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
hPRa 2+hPRa 3
Ready-to-use for IHC (UVLP)
Murine
IgG2a kappa
ASR, IVD
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit