Learn More
Epredia™ Lab Vision™ Progesterone Receptor Rabbit Polyclonal Antibody with BSA and azide
Provide accurate, reproducible results with the Epredia™ Progesterone Receptor, Rabbit Polyclonal Antibody.
Supplier: Epredia™ RB9017P
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: 412-526aa
Immunogen: Recombinant protein encoding human progesterone receptor 412-526aa
Molecular Weight: 81 and 116kDa
Positive Control: T47D cells, Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not tested)
Note:
USA: RUO; Int'l: RUO
Specifications
Progesterone Receptor | |
Polyclonal | |
200µg/mL | |
Rabbit | |
1 mL | |
Cancer and Tumor Biology | |
Human | |
IgG |
Immunohistochemistry (Paraffin) | |
Polyclonal | |
Unconjugated | |
Recombinant protein encoding human progesterone receptor 412-526 aa | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.