Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone, Wettable, Powder, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
57-83-0
C21H30O2
314.47
RJKFOVLPORLFTN-LEKSSAKUSA-N
(1S,3aS,3bS,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
57-83-0
C21H30O2
314.47
RJKFOVLPORLFTN-LEKSSAKUSA-N
(1S,3aS,3bS,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

PR111, 57-83-0, C21H30O2

Spectrum™ Progesterone, Wettable, Powder, USP
Packaging Amber Glass Bottle
Specific Rotation +175° to +183°
Loss on Drying 0.005
Melting Range 126-131°C
Quantity 100 g
CAS Max % 1
Percent Purity 97 to 103%
Grade USP