Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bollé Safety™ Progressif FreeForm Arizona Photochromic Trivex Rx Lens
Prescription lens option for safety glasses.
$359.50
Specifications
Lens Coating | Uncoated |
---|---|
Lens Type | Progressive Photochromatic |
Product Type | Prescription Lens |
Disposable | Reusable |
Lens Material | Plastic |
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
18-072-188
|
Bollé Safety™
PXLPROST02 |
Brown |
Each for $359.50
|
|
|||||
18-072-189
|
Bollé Safety™
PXLPROST03 |
Gray |
Each for $359.50
|
|
|||||
Description
- Lenses made of plastic
- Available in multiple colors
Specifications
Uncoated | |
Prescription Lens | |
Plastic |
Progressive Photochromatic | |
Reusable | |
Not Autoclavable |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title