Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bollé Safety™ Progressif FreeForm+ Colorado Rx Lens
Lens option for prescription safety glasses.
$307.50 - $344.00
Specifications
Lens Coating | Uncoated |
---|---|
Lens Type | Progressive |
Product Type | Prescription Lens |
Disposable | Reusable |
Autoclavable | Not Autoclavable |
Catalog Number | Mfr. No. | Lens Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Material | Price | Quantity & Availability | |||||
18-072-149
|
Bollé Safety™
CRPF1 |
CR39 |
Each for $307.50
|
|
|||||
18-072-168
|
Bollé Safety™
PCPF1 |
Polycarbonate |
Each for $307.50
|
|
|||||
18-072-184
|
Bollé Safety™
PXLPROFT01 |
Trivex |
Each for $344.00
|
|
|||||
Description
- For use with prescription frames
Specifications
Uncoated | |
Prescription Lens | |
Not Autoclavable |
Progressive | |
Reusable |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title