Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proguanil hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mg
100 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 245.0°C to 250.0°C |
Quantity | 25 mg |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Formula Weight | 290.19 |
Percent Purity | 97% |
Chemical Name or Material | Proguanil hydrochloride |
Safety and Handling
Hazard Category | Acute toxicity (category 3) |
Hazard Statement | Toxic if swallowed |
EINECSNumber | 211-283-7 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction