Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prohibitin Mouse anti-Human, Clone: PHB/3229, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30760210UG
Description
Prohibitin Monoclonal specifically detects Prohibitin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
Prohibitin | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
PHB1, prohibitin | |
Recombinant human Prohibitin protein fragment (aa167-261) (exact sequence is proprietary) | |
100 μg | |
Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
5245 | |
Store at 4°C. | |
IgG2a κ |
Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
PHB/3229 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title