Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prohibitin Polyclonal Antibody, Invitrogen™

Catalog No. PIPA124426
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA124426 50 μg
1 options

Catalog No. PIPA124426

Supplier: Thermo Scientific PA124426

Rabbit Polyclonal Antibody

PA1-24426 detects Prohibitin (PHB) in Human samples. PA1-24426 has been successfully used in Western Blot procedures dilute this antibody with 5% skim milk/PBS buffer. PA1-24426 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of of human PHB. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Prohibitin is from an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3′ untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3′ untranslated region may function as a trans-acting regulatory RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prohibitin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene PHB
Gene Accession No. P35232
Gene Alias PHB
Gene Symbols PHB
Host Species Rabbit
Immunogen Synthetic peptide region of of human PHB.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5245
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.