Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS™ ProKlenz RESTORE High Performance Acid Based Cleaner
Click to view available options
Size:
5 gal.
55 gal.
Container Type:
Drum
Pail
Description
ProKlenz RESTORE High Performance Acid Based Cleaner is a globally available, low foaming acid based product for type I rouge removal and all-purpose cleaning. The global acceptability of ProKlenz RESTORE accommodates the pharmaceutical, biotechnology, cosmetic, and nutraceutical industries by removing difficult process residues.
HOW PROKLENZ RESTORE WORKS
ProKlenz RESTORE Cleaner has an acidic pH for the removal of type I rouge and all-purpose cleaning.
WHY PROKLENZ RESTORE?
- Standardization - Globally available product enables harmonized cleaning protocols across sites.
- Sustainability - Free rinsing formula minimizes utility consumption and detergent residue
- Effectiveness - An acidic pH removes type I rouge while a low foaming system supports use in high-impingement systems
- Efficiency - A low use-concentration supports cost-effective and optimized cleaning
- Quality - Manufactured within an FDA registered plant, ISO 9001:2015 and ISO 13485:2016 compliant, to meet the highest standard in manufacturing and quality assurance
Specifications
Specifications
Product Type | Acid Cleaner |
Color-coded | No |
Form | Clear Liquid |
pH | 2.4 (Diluted) |
Scent | Unscented |
Sterility | Non-sterile |
Container Type | Pail |
Size | 5 gal. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction