Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prolactin Rabbit anti-Human, Clone: PRL/4908R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Prolactin
Clone PRL/4908R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121955
View Documents
Novus Biologicals
NBP30724110UG
100 μg
Each for $406.52
Add to cart
 
NB121956
View Documents
Novus Biologicals
NBP30724120UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Prolactin
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
prolactin
Recombinant fragment of human Prolactin protein (around aa 63-201) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
PRL/4908R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction
10 mM PBS with 0.05% BSA
5617
IgG Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit