Learn More
Alfa Aesar™ Prolactin-Releasing Peptide (12-31), Rat
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66882EXD
 View more versions of this product
Catalog No. AAJ66882EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
2272.60
Rat
Solid
RUO
PrRP-20, rat
HPLC
0.5mg
Thr-Pro-Asp-Ile-Asn-Pro-Ala-Trp-Tyr-Thr-Gly-Arg-Gly-Ile-Arg-Pro-Val-Gly-Arg-Phe-NH2
-30° to -10°C
Prolactin-Releasing Peptide (12-31), rat