Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proline, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
147-85-3
C5H9NO2
115.13
ONIBWKKTOPOVIA-UHFFFAOYNA-N
pyrrolidine-2-carboxylic acid
OC(=O)C1CCCN1
147-85-3
C5H9NO2
115.13
ONIBWKKTOPOVIA-UHFFFAOYNA-N
pyrrolidine-2-carboxylic acid
OC(=O)C1CCCN1

P1434, 147-85-3, C5H9NO2

Spectrum™ Proline, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Ignition Residue 0.004
Iron (Fe) 30 ppm
Loss on Drying 0.004
Packaging Poly Pail
Specific Rotation -84.3° to -86.3°
Sulfate 0.0003
Percent Purity 98.5101.5%
Grade USP
Quantity 5 kg