Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Prolyl Oligopeptidase/PREP Human Recombinant Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP2518390.02MG

 View more versions of this product

Catalog No. NBP25839002


Add to cart

Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-710 of Human Prolyl Oligopeptidase/PREP The Recombinant Human Prolyl Oligopeptidase/PREP Protein is derived from E. coli. The Recombinant Human Prolyl Oligopeptidase/PREP Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.25mg/mL
PBS buffer (pH 7.4) containing 30% glycerol, 1mM DTT
TMW: 83.1kDa
0.02mg
dJ355L5.1 (prolyl endopeptidase), EC 3.4.21.26, MGC16060, PE, PEP, Post-proline cleaving enzyme, prolyl endopeptidase, prolyl oligopeptidase
Unlabeled
SDS-Page
5550
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Prolyl Oligopeptidase/PREP
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only and is not approved for use in humans or in clinical diagnosis