Learn More
PROM1 Mouse anti-Human, Clone: 3D4, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation H00008842M10
Catalog No. 89-032-231
Add to cart
Description
Description
Mouse monoclonal antibody raised against a partial recombinant PROM1.
  • High specificity
  • High reactivity
Specifications
Specifications
PROM1
3D4
Mouse monoclonal antibody raised against a partial recombinant PROM1.
NM_006017.2
AC133|CD133|MSTP061|PROML1|RP41
Mouse
IgG1 Kappa
RUO
Primary
8842
ELISA, Immunofluorescence, Western Blot
Unconjugated
In 1x PBS, pH 7.4
Liquid
PROM1
PROM1 (NP_006008.1, 747 a.a. ~ 760 a.a) partial recombinant protein.
100 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)