Learn More
Promega 5X Colorless GoTaq™ Flexi Buffer
5X 5X Colorless GoTaq Flexi Reaction Buffers are Mg-free buffers that allow optimization of Mg concentration in PCR. The green buffer allows direct-to-gel analysis of reaction products without the need to add loading dye.
Supplier:  Promega M8901
Catalog No. PRM8901
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • Mg-Free Buffers for PCR Optimization
  • Use with GoTaq(R) Flexi DNA Polymerase and Mg Solution to optimize magnesium concentration in PCR
  • Available with green dye for direct-to-gel convenience, or in colorless format for applications where absorbance or fluorescence measurements are necessary prior to PCR cleanup
Specifications
Specifications
Colorless
8.5
Liquid
20 mL
5X
Reaction Buffer
25 to 50% Sucrose, pure
−30 to −10°C