Learn More
Promega AKT1 Kinase Enzyme System
Recombinant full-length human AKT1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. AKT1/PKB alpha is a serine/threonine kinase that belongs to the AKT family.
$541.00 - $1054.00
Includes: AKT1 Kinase Enzyme System (V1911)
  • Kinase: AKT1, 10μg (Human, recombinant; amino acids 695–end). MW: ∽85kDa
  • Substrate: Akt (PKB) substrate (CKRPRAASFAE); derived from the N-terminus of GSK3.
  • Reaction Buffer, DTT, MnCl
ADP-Glo™ Kinase Assay and AKT1 Kinase Enzyme System (V1911)
  • 1 x 10μg AKT1 Kinase Enzyme System (V1911) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV1911
View Documents
Promega
V1911
10 μg
Each for $541.00
Add to cart
PRV9061
View Documents
Promega
V9061
1/Ea.
Each for $1,054.00
Add to cart
Description
Description
  • Easily Screen and Profile AKT1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity