Learn More
Promega AMPK (A1/B1/G1) Kinase Enzyme System
Recombinant full-length human AMPK (combination of A1/B1/G1 subunits) was expressed by baculovirus in Sf9 insect cells using C-terminal His tags. AMPK is an important energy-sensing enzyme that monitors cellular energy status.
$1148.00 - $1531.00
Specifications
Format Fluid
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV1921
View Documents Promotion Details
Promega
V1921
10 μg
Each for $1,148.00
Add to cart
PRV9021
View Documents Promotion Details
Promega
V9021
1/Ea.
Each for $1,531.00
Add to cart
Description
Description
  • Easily Screen and Profile AMPK (A1/B1/G1) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
Fluid