Learn More
Promega Bovine Serum Albumin, Acetylated
Used as an enzyme stabilizer or as a carrier protein. Acetylated to be nuclease-free.
Supplier: Promega R9461
Description
- Quality tested and certified free of DNase and RNase activity
Specifications
1μg/μL | |
400μL | |
−30 to −10°C |
BSA Acetylated | |
Bovine Serum | |
Liquid |