Learn More
Promega c-MER Kinase Enzyme System
Recombinant human c-MER (amino acids 578-872) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. c-MER is a tyrosine kinase and a member of the MER/AXL/TYRO3 receptor kinase family.
$540.50 - $1070.00
Specifications
Storage Requirements <−65°C
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity & Availability  
PRV3541
View Documents Promotion Details
Promega
V3541
Involved in engulfment and clearance of Apoptotic Cells 10μg
Each for $540.50
Add to cart
PRV9561
View Documents Promotion Details
Promega
V9561
Involved in engulfment and clearance of Apoptotic Cells, used to monitor the activity of virtually any ADP-generating Enzyme 1/ea
Each for $1,070.00
Add to cart
Description
Description
  • Easily Screen and Profile c-MER Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
<−65°C