Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega CAMK2gamma Kinase Enzyme System

Recombinant human CAMK2 gamma (C-terminal truncation) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. CAMK2 gamma is a member of the CAMKII family.

$540.50 - $1070.00

Specifications

Format Fluid
Molecular Weight 60
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Quantity Price Quantity & Availability  
PRV3531
View Documents
Promega
V3531
Implicated in diverse effects of hormones and neurotransmitters 10 μg
Each for $540.50
Add to cart
 
PRV9201
View Documents
Promega
V9201
Implicated in diverse effects of hormones and neurotransmitters and both primary screening as well as Kinase selectivity profiling 1/Ea.
Each for $1,070.00
Add to cart
 
Description

Description

  • Easily Screen and Profile CAMK2γ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications

Specifications

Fluid
60
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit