Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega cAMP-Dependent Protein Kinase, Catalytic Subunit
May be used to phosphorylate target proteins or for in vitro enzymological studies of neural and hormonal signal transduction
Supplier: Promega ZV5161
Description
- 40kDa protein purified from a recombinant E. coli strain expressing the catalytic subunit of bovine PKA
- Purity: 90%
- Intracellular targets include ion channels, transcriptional activator proteins, and regulatory enzymes of glycogen metabolism
- Unit Definition: One unit is defined as the amount of enzyme required to incorporate 1pmol of phosphate into casein in one minute at 30°C
- Storage Buffer: 350mM potassium phosphate (pH 6.8), 0.1mM DTT
Specifications
1.5 to 3 mg/mL | |
cAMP-dependent Protein Kinase |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction