Learn More
Promega Caspase-Glo™ 2 Assay System
A homogeneous, luminescent assay that measures caspase-2 activity in biochemical reactions with purified caspase-2
Manufacturer:  Promega G0940
Catalog No. PRG0940
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Flexible, simplified method provides greater sensitivity, faster results
  • Broad dynamic range
  • Demonstrates Z-factor value of 0.85 in 384-well plates using 0.05U/ml of enzyme
  • Increased accuracy, batch-processing capability
  • Includes: Caspase-Glo 2 Buffer, Z-VDVAD-Glo™ Substrate, Luciferin Detection Reagent
  • Suitable for biochemical screens for Caspase -2 inhibitors
Applications:
Caspase-2 inhibitor studies
Specifications
Specifications
Ideal for biochemical screens for Caspase -2 inhibitors
Fluid
1 x 10mL Caspase-Glo™ 2 Buffer, 1 x 100μL Z-VDVAD-Glo™ substrate, Luciferin Detection Reagent
10 mL
7.2