Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega CHK1 Kinase Enzyme System

Recombinant full-length human CHK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CHK1 was originally identified in fission yeast to play a role in activation of the DNA damage checkpoint in the G2 phase of the cell cycle.

$544.00 - $1070.00

Specifications

Format Fluid
Molecular Weight 82
View More Specs

Includes:
  • Kinase: CHK1, 10μg (Human, recombinant full length). MW: ∽82kDa
  • Substrate: CHKtide (KKKVSRSGLYRSPSMPENLNRPR); derived from the human CDC25C protein isoform A (amino acid 205-225)
  • Others: Reaction Buffer, DTT

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV1941
View Documents
Promega
V1941
10 μg
Each for $544.00
Add to cart
 
PRV9241
View Documents
Promega
V9241
1/Ea.
Each for $1,070.00
Add to cart
 
Description

Description

  • Easily Screen and Profile CHK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications

Specifications

Fluid
82
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit