Learn More
Promega Chloramphenicol Acetyltransferase
Manufacturer:  Promega E1051
Catalog No. PR-E1051
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Chloramphenicol Acetyltransferase
Liquid
Colorless
Catalyzes the transfer of an Acetyl group from Acetyl-CoA to the 3-hydroxy position of Chloramphenicol
100 U
10 to 14 U/μL
7.8
Reporter lysis 5X Buffer