Learn More
Promega CSK Kinase Enzyme System
Recombinant full-length human CSK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CSK has been shown to downregulate the tyrosine kinase activity of c-src through tyrosine phosphorylation of the c-src carboxy terminus.
$540.50 - $1070.00
Includes:
  • 1 x 10μg CSK Kinase Enzyme System (V2981) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV2981
View Documents Promotion Details
Promega
V2981
10 μg
Each for $540.50
Add to cart
PRV9251
View Documents Promotion Details
Promega
V9251
1/Ea.
Each for $1,070.00
Add to cart
Description
Description
  • Easily Screen and Profile CSK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity