Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega DeadEnd™ Colorimetric TUNEL System

A modified TUNEL Assay designed for simple, rapid detection of apoptotic cells in situ at the single-cell level. Measures nuclear DNA fragmentation.

$542.50 - $777.00

Specifications

Detection Method Colorimetric
Format Fluid
For Use With (Application) Detection of apoptotic cells in situ at the single-cell level
pH 6.8
Storage Requirements COMPONENTS AT DIFFERENT TEMPS
View More Specs

Includes: DAB substrate and H2O2 plus plastic coverslips for easy sample handling

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PR-G7360
View Documents
Promega
G7360
20 Reactions
Each for $542.50
Add to cart
 
PR-G7130
View Documents
Promega
G7130
40 Reactions
Each for $777.00
Add to cart
 
Description

Description

  • A Modified TUNEL Assay for Detection of Apoptotic Cells In Situ
  • Detects apoptosis in tissue sections or cell cultures
Specifications

Specifications

Colorimetric
Detection of apoptotic cells in situ at the single-cell level
COMPONENTS AT DIFFERENT TEMPS
Fluid
6.8
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit