Learn More
Promega EnduRen™ Live Cell Substrate
An automation-friendly, highly sensitive substrate for Renilla luciferase-based gene reporter and BRET applications.
$318.50 - $13910.00
Specifications
Content And Storage -30°C to -10°C
View More Specs
Products 3
Catalog Number Mfr. No. Includes Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Includes Quantity Price Quantity & Availability  
PR-E6482
View Documents
Promega
E6482
1 x 3.4mg EnduRen™ live Cell substrate 3.4 mg
Each for $1,999.00
Add to Cart
PR-E6481
View Documents
Promega
E6481
1 x 0.34mg EnduRen™ live Cell substrate 0.34 mg
Each for $318.50
Add to Cart
PRE6485
View Documents
Promega
E6485
1 x 34mg EnduRen™ live Cell substrate 34 mg
Each for $13,910.00
Add to Cart
Description
Description
  • Live-Cell Detection of Renilla Luminescence for up to 24 Hours
  • Engineered coelenterazine for live-cell reporter gene, BRET and RNAi assays
  • Automation-friendly and highly sensitive
Specifications
Specifications
-30°C to -10°C