Learn More
Promega EPHA1 Kinase Enzyme System
Recombinant mouse EPHA1 (amino acids 569-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EPHA1 is a member of the Eph family of receptor tyrosine kinases that have been implicated in mediating developmental events.
$540.50 - $1070.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV3561
View Documents
Promega
V3561
10 μg
Each for $540.50
Add to cart
PRV9271
View Documents
Promega
V9271
Each
Each for $1,070.00
Add to cart
Description
Description
  • Easily Screen and Profile EPHA1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity