Learn More
Promega FGFR1 Kinase Enzyme System
Recombinant human FGFR1 (amino acids 399-822) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FGFR1 (also known as FLT2) is a member of the Fibroblast Growth Factor Receptor family.
$540.50 - $1070.00
Specifications
Storage Requirements <−65°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV2991
View Documents Promotion Details
Promega
V2991
10μg
Each for $540.50
Add to cart
PRV9321
View Documents Promotion Details
Promega
V9321
1/ea
Each for $1,070.00
Add to cart
Description
Description
  • Easily Screen and Profile FGFR1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
<−65°C