Learn More
Promega FYN A Kinase Enzyme System
Full-length recombinant human FYN A was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FYN A (Fyn isoform A) is a member of the SRC tyrosine kinase oncogene family showing high homology to YES1, FGR and SRC.
$537.50 - $1060.00
Includes: FYN A Kinase Enzyme System (V3571)
  • Kinase: FYN A, 10μg (Human, recombinant full-length). MW: ∽85kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide.
  • Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9341)
  • 1 x 10μg EGFR Kinase Enzyme System (V3571) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV3571
View Documents
Promega
V3571
10 μg
Each for $537.50
Add to cart
PRV9341
View Documents
Promega
V9341
1/Ea.
Each for $1,060.00
Add to cart
Description
Description
  • Easily Screen and Profile FYN A Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity