Learn More
Promega GloMax 20/20 Luminometry System
Touchscreen interface simplifies operation
Specifications
Holds | 1.5mL microcentrifuge tubes and 35mm Petri dishes |
---|---|
Range | >8 decades |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
PRE5321
|
Promega
E5321 |
With single autoinjector |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
PRE5331
|
Promega
E5331 |
With dual autoinjectors |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Key Features
- Units are DLReady certified for optimum results using Promega's Dual-Luciferase Reporter Assays
- Promega protocols are preloaded into the software for easy start-up
- Optional automatic injectors enable measurement of flash reactions
- Proprietary circuitry and advanced photon-counting photomultiplier tube (PMT) provide superior signal-to-noise ratios
- Extended dynamic range permits measurement of both dim and bright samples without dilution and minimizes the chance of detector saturation
Additional Features
- Menu-driven prompts guide user through protocol setup
- Up to 18 protocols can be stored for future use
- Unit may be set to start measurement immediately after lid is closed
- Connections, priming and flushing are simplified because optional injectors fit next to the detection module
- Completely visible plumbing allows inspection of all tubing and tips during flushing and priming
- Spreadsheet interface software allows data to be exported to Excel
- Software can be programmed to subtract background from all measurements, calculate the average and standard deviation of replicate samples and report kinetic measurements
Order Info
Models are available with a choice of zero, one or two reagent injectors.
Specifications
1.5mL microcentrifuge tubes and 35mm Petri dishes | |
>8 decades |