Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega GSK3α Kinase Enzyme System
Recombinant full-length human GSK3 alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. GSK3 alpha is a multifunctional protein serine kinase implicated in the control of several regulatory proteins.
$748.00 - $1421.00
Specifications
Molecular Weight (g/mol) | 81 |
---|---|
Form | Fluid |
Includes:
- 1 x 10μg GSK3α Kinase Enzyme System (V3051) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
Description
- Easily Screen and Profile GSK3α Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
81 | |
Fluid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title