Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HaloTag™ Alexa Fluor™ 660 Fluorescent Ligand

The HaloTag™ Technology enables covalent protein labeling and immobilization in vivo and in vitro.

$647.50 - $1048.00

Specifications

Concentration 3.5 mM
For Use With (Application) Protein localization, Protein trafficking, Protein capture, cell-to -gel analysis, Protein interaction analysis
Product Type HaloTag™ Fluorescent Ligand
Sufficient For For cell-surface labeling
Content And Storage -30°C to -10°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRG8471
View Documents
Promega
G8471
30 μL
Each for $1,048.00
Add to Cart
 
PRG8472
View Documents
Promega
G8472
15 μL
Each for $647.50
Add to Cart
 
Description

Description

  • Covalently Attach Fluorescent Labels or Affinity Tags to Membrane or Intracellular Proteins
  • Cell permeant TMR, Oregon Green(R), diAcFAM and Coumarin ligands readily cross the cell membrane for labeling intracellular proteins
  • Cell impermeant tags include Alexa Fluor(R) 488 and 660 for quick cell surface labeling
Specifications

Specifications

3.5 mM
HaloTag™ Fluorescent Ligand
-30°C to -10°C
Protein localization, Protein trafficking, Protein capture, cell-to -gel analysis, Protein interaction analysis
For cell-surface labeling
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit