Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HER2 Kinase Enzyme System

Recombinant human HER2 (amino acids 676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HER2 gene encodes a cell-surface glycoprotein tyrosine kinase receptor with extensive homology to epidermal growth factor receptor.

$600.00 - $1070.00

Specifications

Format Fluid
For Use With (Application) Cellular transformation and tumorigenesis
Molecular Weight 116
Storage Requirements -70°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV3891
View Documents Promotion Details
Promega
V3891
10μg
Each for $600.00
Add to cart
 
PRV9381
View Documents Promotion Details
Promega
V9381
1000 Assays
Each for $1,070.00
Add to cart
 
Description

Description

  • Easily Screen and Profile HER2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications

Specifications

Fluid
Cellular transformation and tumorigenesis
116
-70°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit