Learn More
Promega HER4 Kinase Enzyme System
Recombinant human HER4 (amino acids 682-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. HER4 or ERBB4 is a transmembrane receptor tyrosine kinase that belongs to the epidermal growth factor receptor family.
$540.50 - $1070.00
Includes: HER4 Kinase Enzyme System (V3101)
  • Kinase: HER4, 10μg (Human, recombinant; amino acids 682–993). MW: ∽57kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide.
  • Others: Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and HER4 Kinase Enzyme System (V9391)
  • 1 x 10μg EGFR Kinase Enzyme System (V3101) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV3101
View Documents
Promega
V3101
10 μg
Each for $540.50
Add to cart
PRV9391
View Documents
Promega
V9391
1/Ea.
Each for $1,070.00
Add to cart
Description
Description
  • Easily Screen and Profile HER4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity