Learn More
Promega HisLink 96 Protein Purification System
HisLink Protein Purification Systems are designed for purification of polyhistidine (His)-tagged or HQ-tagged proteins directly from culture medium containing bacterial cells expressing the tagged protein of interest.
Specifications
Product Type | HisLink™ 96 Protein Purification System |
---|---|
For Use With (Application) | Quick and robust method of purifying multiple Polyhistidine-or HQ-tagged expressed Proteins from E. coli using a vacuum-based method |
Storage Requirements | 2 to 10°C |
Requires: 96-place vacuum manifold
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PRV3681
|
Promega
V3681 |
5 x 96 Tests |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
PRV3680
|
Promega
V3680 |
1 x 96 Tests |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Capture His- or HQ-Tagged Protein Cell Lysates by Centrifugation or 96-Well Plate
- Tagged proteins purified directly from cell lysates
- Purify manually by batch, using a vacuum manifold, liquid chromatography or liquid handling platform
- Purify by centrifugation or vacuum method
Specifications
HisLink™ 96 Protein Purification System | |
Quick and robust method of purifying multiple Polyhistidine-or HQ-tagged expressed Proteins from E. coli using a vacuum-based method | |
2 to 10°C |
Patent Pending.
This product is licensed for use under U.S. Pat. No. 6,174,704.
Certain applications of this product are covered by patents issued to parties other than Promega and applicable in certain countries. Because purchase of this product does not include a license to perform any of these patented applications, users of this product may be required to obtain a patent license depending upon the particular application and country in which the product is used.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.