Learn More
Promega JAK3 Kinase Enzyme System
Recombinant human JAK3 (amino acids 781-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. JAK3 is a member of the JAK family of tyrosine kinases involved in cytokine receptor-mediated intracellular signal transduction.
$536.00 - $1060.00
Includes: JAK3 Kinase Enzyme System (V3701)
  • 10μg of JAK3 Kinase (Human, recombinant; amino acids 781–end)
  • Poly (4:1 Glu4,Tyr1) peptide substrate
  • Reaction buffer
  • DTT
ADP-Glo™ Kinase Assay and JAK3 Kinase Enzyme System (V9441)
  • 1 x 10μg JAK3 Kinase Enzyme System (V3831) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV3701
View Documents
Promega
V3701
10 μg
Each for $536.00
Add to cart
PRV9441
View Documents
Promega
V9441
1/Ea.
Each for $1,060.00
Add to cart
Description
Description
  • Easily Screen and Profile JAK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity